Would you like to log in again?
SensoLyte® Cell Viability and Proliferation Assay Kits, AnaSpec
Supplier: AnaSpec
AS-71301 AS-71300
103008-892EA 483.77 USD InStock
103008-892 103010-254
SensoLyte® Cell Viability and Proliferation Assay Kits, AnaSpec
Assays Enzyme Assays
Lactate dehydrogenase (LDH) exists ubiquitously in the cytoplasm of living mammalian cells and the measurement of cytoplasmic LDH activity is a well-accepted assay to quantify living cell numbers and indicate cell viablity.
The SensoLyte® Cell Viability and Proliferation Assay Kit provides researchers with a convenient one-solution and one-step assay to count living cells in a culture and continuously monitor cell proliferation over time by measuring cytoplasmic LDH activity. In this kit, resazurin is used as a sensitive fluorogenic indicator. Resazurin is converted to the strongly fluorescent resorufin (Ex/Em=560nm/590 nm) by cytoplasmic LDH. The kit can detect as few as 48 living cells with a linear range up to 5X104 cells (R2=0.99). This kit can also be used for high throughput screening of cell proliferation or measuring cytotoxicity effect of a variety of compounds. 384-well or 1536-well format can be used with minor modifications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty