Would you like to log in again?
SensoLyte® Cell Cytotoxicity Assay Kits, AnaSpec
Supplier: AnaSpec
AS-71303 AS-71302
103010-256EA 483.77 USD InStock
103010-256 103010-258
SensoLyte® Cell Cytotoxicity Assay Kits, AnaSpec
Assays Enzyme Assays
The damage of cell membrane leads to the release of cytoplasmic enzymes
The SensoLyte® Cell Cytotoxicity Assay Kit uses resazurin as a sensitive fluorogenic indicator (Ex/Em=560 nm/590 nm upon conversion) to measure LDH activity. The assay can be performed in a mixed population of damaged and viable cells, but it only measures the LDH released from damaged cells. The cytoplasmic LDH in living cells produces little signals under assay condition. There is no need for extra steps to separate living cells and supernatant. The fluorescent signal is proportional to the number of damaged cells (up to 2.5 x 104 cell, r2>0.95) with the detection limit reaching 100 dead cells. The kit is suitable for high throughput screening of cytotoxicity of a variety of compounds. 384-well or 1536-well format can be used with minor modifications.
The measurement of released cytoplasmic lactate dehydrogenase (LDH) is a well-accepted assay to estimate cell membrane integrity and quantify cytotoxicity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty