Would you like to log in again?
SensoLyte® Plus 520 MMP-13 Assay Kit (Fluorimetric and Enhanced Selectivity), AnaSpec Inc.
Supplier: AnaSpec
AS-72019
103010-292EA 969 USD InStock
103010-292
SensoLyte® Plus 520 MMP-13 Assay Kit (Fluorimetric and Enhanced Selectivity), AnaSpec Inc.
Assays Enzyme Assays
The SensoLyte® Plus 520 MMP-13 Assay Kit is designed for specifically detecting MMP-13 activity in biological samples which may contain multiple MMPs, such as culture medium, serum, plasma, synovial fluid, and tissue homogenate. A specific anti-MMP-13 monoclonal antibody is used in combination with a MMP fluorogenic substrate, 5-FAM/QXL®520 FRET peptide. The fluorescence signal is monitored at Ex/Em=490 nm/520 nm upon MMP-13-induced cleavage of the FRET substrate. Ample materials are provided to perform 96 assays in a 96-well format.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty