Would you like to log in again?
SensoLyte® 520 MMP - 10 Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72024
103010-302EA 809.69 USD InStock
103010-302
SensoLyte® 520 MMP - 10 Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
MMP-10 (stromelysin 2) cleaves extracellular matrix proteins, but is unable to cleave the triple-helical fibrillar collagens
The SensoLyte® 520 MMP-10 Assay Kit uses a 5-FAM (fluorophore) and QXL®520 (quencher) labeled FRET peptide substrates for continuous measurement of the enzyme activities. In an intact FRET peptide, the fluorescence of 5-FAM is quenched by QXL® 520. Upon the cleavage of the FRET peptide by MMP-10, the fluorescence of 5-FAM is recovered, and can be continuously monitored at excitation/emission = 490 nm/520 nm. With superior fluorescence quantum yield and longer emission wavelength, 5-FAM/QXL®520 based FRET peptide is less interfered by the autofluorescence of test compounds and cellular components and provides better assay sensitivity. The assays are performed in a convenient 96-well or 384-well microplate format.
This enzyme is involved in several pathological conditions, such as cancer, arthritis and wound healing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty