Would you like to log in again?
SensoLyte® 390 Renin Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72039
103010-340EA 903.23 USD InStock
103010-340
SensoLyte® 390 Renin Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
Renin, a highly specific aspartyl protease, cleaves angiotensinogen, to yield angiotensin I, which is further converted into angiotensin II by ACE (Angiotensin Converting Enzyme)
The SensoLyte® 390 Renin Assay Kit provides a convenient assay for high throughput screening of renin inhibitors and for continuous assay of renin activity using a Mc-Ala/Dnp fluorescence resonance energy transfer (FRET) peptide. In the FRET peptide the fluorescence of Mc-Ala is quenched by Dnp. Upon cleavage into two separate fragments by renin, the fluorescence of Mc-Ala is recovered, and can be monitored at excitation/emission = 330/390 nm. This assay is about four fold more sensitive than an EDANS/DABCYL-based assay and can detect 8 ng/mL of renin.
Angiotensin II constricts blood vessels leading to increased blood pressure. It also increases the secretion of ADH and aldosterone, and stimulates the hypothalamus to activate the thirst reflex. Since an overactive renin-angiotensin system leads to hypertension, renin is an attractive target for the treatment of this disease.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty