Would you like to log in again?
Human Recombinant Renin (from HEK293 Cells)
Supplier: AnaSpec
AS-72041
103010-344EA 188.54 USD InStock
103010-344
Human Recombinant Renin (from HEK293 Cells)
Proteins and Peptides
Renin, a highly specific aspartyl protease, cleaves angiotensinogen, produced in the liver, to yield angiotensin I, which is further converted into angiotensin II by ACE (Angiotensin Converting Enzyme). Angiotensin II constricts blood vessels, leading to increased blood pressure. It also increases the secretion of ADH and aldosterone, and stimulates the hypothalamus to activate the thirst reflex. Since an overactive renin-angiotensin system leads to hypertension, renin is proposed as a therapeutic target for this disease
The recombinant human pro-renin was expressed in HEK cells and converted to the active renin. After affinity chromatography, the active enzyme has a purity of >99% by SDS-PAGE. The molecular mass of active human renin is approximately 40 kDa.
The activity of enzyme can be measured in FRET-based assays
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty