Would you like to log in again?
AnaTag™ HiLyte™ Fluor 750 Microscale Protein Labeling Kit *Ultra Convenient*, Anaspec
Supplier: AnaSpec
AS-72044
103010-348EA 418 USD InStock
103010-348
AnaTag™ HiLyte™ Fluor 750 Microscale Protein Labeling Kit *Ultra Convenient*, Anaspec
Protein Linking and Labeling Systems
HiLyte™ Fluor 750 acid, SE is the longest-wavelength amine-reactive HiLyte™ Fluor dye currently available. Its fluorescence emission maximum at 778 nm is well separated from commonly used far-red fluorophores such as HiLyte™ Fluor 647, HiLyte™ Fluor 680 or allophycocyanin (APC), facilitating multicolor analysis.
With a peak excitation at ~754 nm, conjugates of HiLyte™ Fluor 750 dyes are well excited by a xenon-arc lamp or dye-pumped lasers operating in the 720–750 nm range. The HiLyte™ Fluor 750-protein conjugates are suitable for immunofluorescent staining, in situ hybridization, flow cytometry and other biological applications. The kit has all essential components for performing the conjugation reaction and for purifying the HiLyte™ Fluor 750-protein conjugates. The kit is able to perform three conjugation reactions with each reaction labeling up to 200 µg IgG. Kit Size: 3 labeling reactions (3 x 200 µg protein)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty