Would you like to log in again?
AnaTag™ HiLyte™ Fluor 555 Protein Labeling Kit *Ultra Convenient* [3 labeling reactions (3 x 5 mg protein)], Anaspec
Supplier: AnaSpec
AS-72045
103010-350EA 663.54 USD InStock
103010-350
AnaTag™ HiLyte™ Fluor 555 Protein Labeling Kit *Ultra Convenient* [3 labeling reactions (3 x 5 mg protein)], Anaspec
Protein Linking and Labeling Systems
HiLyte™ Fluor 555 acid, SE is an excellent amine-reactive fluorescent labeling dye that generates the protein conjugates that are only slightly red-shifted compared to those of Cy™ 3 dyes, resulting in an optimal match to filters designed for CyTM dyes. The HiLyte™ Fluor 555-protein conjugates are very stable and suitable for immunofluorescent staining, in situ hybridization, flow cytometry and other biological applications. Additionally, our in-house data indicated that HiLyte™ Fluor 555 is superior to CyTM 3 in fluorescence polarization-based assays. The kit has all essential components for performing the conjugation reaction and for purifying the HiLyte™ Fluor 555-protein conjugate. The entire process takes only half an hour. The kit is able to perform three conjugation reactions with each reaction labeling up to 5 mg IgG. Kit Size: 3 labeling reactions (3 x 5 mg protein)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty