Would you like to log in again?
AnaTag™ HiLyte™ Fluor 488 Microscale Protein Labeling Kit, Anaspec
Supplier: AnaSpec
AS-72048-
103010-356EA 418 USD InStock
103010-356
AnaTag™ HiLyte™ Fluor 488 Microscale Protein Labeling Kit, Anaspec
Protein Linking and Labeling Systems
HiLyte™ Fluor 488, SE is an excellent amine-reactive fluorescent labeling dye for generating protein conjugates. The spectrum of HiLyte™ Fluor488 is similar to fluorescein (FITC), resulting in an optimal match to filters designed for fluorescein.
HiLyte™ Fluor 488 is more photostable than FITC, providing researchers with additional time for capturing image. Its fluorescence can be detected at the emission wavelength of 523±10 nm when excited at 499±10 nm. The HiLyte™ Fluor 488-protein conjugates are very stable and suitable for immunofluorescent staining, in situ hybridization, flow cytometry and other biological applications. The kit has all essential components for performing the conjugation reaction and for purifying the HiLyte™ Fluor 488-protein conjugate. The entire process takes only half an hour. The kit is able to perform three conjugation reactions with each reaction labeling up to 200 µg IgG. Kit Size: 3 labeling reactions (3 x 200 µg protein)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty