Would you like to log in again?
AnaTag™ HiLyte™ Fluor 647 Protein Labeling Kit, Ultra Convenient [3 labeling reactions], AnaSpec
Supplier: AnaSpec
AS-72049
103010-358EA 663.54 USD InStock
103010-358
AnaTag™ HiLyte™ Fluor 647 Protein Labeling Kit, Ultra Convenient [3 labeling reactions], AnaSpec
Assays Enzyme Assays
HiLyte™ Fluor 647 acid, SE (Ex/Em=647 nm/679 nm) is an excellent fluorescent labeling dye that generates the protein conjugates that are slightly red-shifted compared to those of Cy® 5 dyes, resulting in an optimal match to filters designed for Cy®5 dyes
The HiLyte™ Fluor 647-protein conjugates are suitable for immunofluorescent staining, in situ hybridization, flow cytometry and other biological applications. Additionally, our in-house data indicated that HiLyte™ Fluor 647 is superior to Cy™5 in fluorescence polarization-based assays. The kit has all essential components for performing the conjugation reaction and for purifying the HiLyte™ Fluor 647-protein conjugate. The entire process takes only half an hour. The kit is able to perform three conjugation reactions with each reaction labeling up to 5 mg IgG. Kit Size: 3 labeling reactions (3 x 5 mg protein).
However, total fluorescence of the secondary antibody conjugates of HiLyte™ Fluor 647 dyes is significantly higher than that of Cy®5 conjugates supplied by other companies. Also, unlike Cy®5 dyes, HiLyte™ Fluor 647 dyes have very little change in absorption or fluorescence spectra when conjugated to most proteins, oligonucleotides and nucleic acids, thus yielding greater total fluorescence at the same degree of substitution.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty