Would you like to log in again?
AnaTag™ AMCA - X Microscale Protein Labeling Kit *Ultra Convenient*, Anaspec Inc.
Supplier: AnaSpec
AS-72056
103010-368EA 394.62 USD InStock
103010-368
AnaTag™ AMCA - X Microscale Protein Labeling Kit *Ultra Convenient*, Anaspec Inc.
Protein Linking and Labeling Systems
AMCA-X, SE is one of the most popular blue fluorescent tagging molecules. It is widely used to label antibodies, proteins and small drug molecules. AMCA-X succinimidyl ester contains a seven-atom aminohexanoyl spacer between the fluorophore and the reactive group.
This spacer separates the fluorophore from the biomolecule to which it is conjugated, potentially reducing the quenching that typically occurs upon conjugation. AMCA fluorescence can be detected at the emission wavelength of 442±10 nm when excited at 353±10 nm. The AMCA-protein conjugates are suitable for immunofluorescent staining, in situ hybridization, flow cytometry and other biological applications. The kit has all essential components for performing the conjugation reaction and for purifying the AMCA-protein conjugates. The kit is able to perform three conjugation reactions with each reaction labeling up to 200 µg IgG. Kit Size: 3 labeling reactions (3 x 200 µg protein)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty