Would you like to log in again?
AnaTag™ 5 - FITC Protein Labeling Kit *Ultra Convenient*, Anaspec
Supplier: AnaSpec
AS-72059
103010-374EA 545.15 USD InStock
103010-374
AnaTag™ 5 - FITC Protein Labeling Kit *Ultra Convenient*, Anaspec
Protein Linking and Labeling Systems
Fluorescein isothiocyanate (FITC) remains one of the most popular fluorescent labeling reagents, probably due to the low cost of the material. 5-FITC and 6-FITC have very similar absorption and fluorescence spectra. However, the isomers may differ in their binding and reactivity to proteins, and the conjugates may elute under different chromatographic conditions or migrate differently in electrophoretic gels.
5-FITC is more widely used than 6-FITC isomer for protein labeling. FITC-protein conjugates are suitable for immunofluorescent staining, in situ hybridization, flow cytometry and other biological applications. The kit has all the essential components for performing the conjugation reaction and for purifying the FITC-protein conjugates. The kit is good for three conjugation reactions with each reaction labeling up to 5 mg IgG. Kit Size: 3 labeling reactions (3 x 5 mg protein)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty