Would you like to log in again?
SensoLyte® 440 West Nile Virus Protease Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72079
103010-398EA 907.62 USD InStock
103010-398
SensoLyte® 440 West Nile Virus Protease Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
West Nile virus (WNV) causes severe neurological disease and fatalities in both human and animal hosts
The SensoLyte® 440 West Nile Virus Protease Assay Kit provides a convenient homogenous assay for high throughput screening of WNV protease inhibitors and for continuous assay of NS3 protease activity using a fluorogenic Pyr-RTKR-AMC peptide. Upon NS3 protease cleavage, Pyr-RTKR-AMC generates the AMC fluorophore that has bright blue fluorescence and can be detected at excitation/emission=354 nm/442 nm.
There is currently no effective vaccine or antiviral drug to protect against WNV infection. The West Nile virus NS3 protease, essential for the proteolytic processing of the viral polyprotein precursor, is a promising target for the development of anti-WNV drugs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty