Would you like to log in again?
SensoLyte® 570 West Nile Virus Protease Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72080
103010-400EA 907.62 USD InStock
103010-400
SensoLyte® 570 West Nile Virus Protease Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
West Nile virus (WNV) causes severe neurological disease and fatalities in both human and animal hosts
The SensoLyte® 570 West Nile Virus Protease Assay Kit provides a convenient homogeneous assay for high throughput screening of WNV protease inhibitors and for continuous assay of NS3 protease activity using QXLTM570/5-TAMRA FRET substrate. Upon cleavage into two separate fragments by the WNV NS3 protease, the fluorescence of 5-TAMRA can be monitored at excitation/emission = 540 nm/575 nm.
There is currently no effective vaccine or antiviral drug to protect against WNV infection. West Nile virus NS3 protease, essential for the proteolytic processing of the viral polyprotein precursor, is a promising target for the development of anti-WNV drugs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty