Would you like to log in again?
SensoLyte® 490 HCV Protease Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-71126 AS-72087
103010-146EA 1175.08 USD InStock
103010-146 103010-406
SensoLyte® 490 HCV Protease Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
HCV protease is identified as an important drug-screening target.
The SensoLyte® 490 HCV Protease Assay Kit uses an optimized FRET peptide substrate for the continuous measurement of HCV NS3/4a activity. This FRET substrate is cleaved specifically by HCV NS3/4a protease thereby liberating the C-terminal peptide-fluorophore fragment from the proximity quenching effect of the dark quencher, resulting in a more than 10-fold increase in fluorescence. This assay can detect HCV NS3/4a protease activity at 1 pmole.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty