Would you like to log in again?
SensoLyte® Generic MMP Assay Kit (Colorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72095
103010-412EA 673.77 USD InStock
103010-412
SensoLyte® Generic MMP Assay Kit (Colorimetric), AnaSpec Inc.
Assays Enzyme Assays
Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated proteins capable of digesting extracellular matrix components
The SensoLyte® Generic MMP Assay Kit can be used to detect the activity of a variety of MMPs, including MMP-1, 2, 3, 7, 8, 9, 12, 13, and 14 or for high throughput screening of MMPs’ inducers and inhibitors. The substrate is a thiopeptolide that is cleaved by the MMPs, releasing a sulfhydryl group, which can be detected with a color-developing thiol-reactive agent, 4,4’-dithiodipyridine or Ellman’s Reagent at 412 nm using a microplate reader. The assays are performed in a convenient 96-well microplate format.
The importance of MMPs in tumor development and invasion as well as other diseases is well known.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty