Would you like to log in again?
SensoLyte® 570 Generic MMP Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72101
103010-424EA 850.62 USD InStock
103010-424
SensoLyte® 570 Generic MMP Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated proteins capable of digesting extracellular matrix components
The SensoLyte® 570 Generic MMP Assay Kit is optimized to detect the activity of a variety of MMPs, including MMP-1, 2, 7, 8, 9, 10, 13, and 14. This kit is ideal for detecting generic MMP activity in biological samples or for high throughput screening of MMP inducers and inhibitors using purified MMPs. This kit uses a 5-TAMRA (fluorophore) and QXL® 570 (quencher) labeled FRET peptide substrate for continuous measurement of the enzyme activities. In an intact FRET peptide, the fluorescence of 5-TAMRA is quenched by QXL® 570. Upon the cleavage of the FRET peptide by MMPs, the fluorescence of 5-TAMRA is recovered, and can be continuously monitored at excitation/emission = 540 nm/575 nm. 5-TAMRA/QXL® 570 based FRET peptide is an ideal substrate for screening MMPs inhibitors since it is less interfered by the autofluorescence of test compounds due to longer emission wavelength. The assays are performed in a convenient 96-well format.
The importance of MMPs in tumor development and invasion as well as other diseases is well known.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty