Would you like to log in again?
AnaTag™ APC Labeling Kit, AnaSpec
Supplier: AnaSpec
AS-72111
103010-444EA 925.15 USD InStock
103010-444
AnaTag™ APC Labeling Kit, AnaSpec
Assays Enzyme Assays
The AnaTag™ APC Labeling Kit is optimized for use in the conjugation of allophycocyanin (APC) to antibodies. APC is an ultra-sensitive, near-infrared fluorescent tracer with a high quantum yield (Ex/Em = 650 nm/660 nm). APC- labeled antibodies are used in applications such as flow cytometry and immunofluorescence. The AnaTagTM APC Labeling Kit contains SH-reactive APC. SMCC modified allophycocyanin reacts with the thiol groups of target antibody without the need for additional activation, thus simplifying the conjugation protocol. AnaTagTM APC Labeling Kit contains a chemically cross-linked APC (CL-APC) that is much more stable than the native APC, but still retains the original spectroscopic properties. The amount of APC supplied in this kit is sufficient for labeling up to 1 mg of antibody. The kit provides all reagents, purification columns and a detailed step-by-step protocol. Cross-linked and SMCC-activated APC are also available as separate products.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty