Would you like to log in again?
SensoLyte® Luminescent Alkaline Phosphatase Assay Kit Luminometric
Supplier: AnaSpec
AS-72122
103010-452EA 663.54 USD InStock
103010-452
SensoLyte® Luminescent Alkaline Phosphatase Assay Kit Luminometric
Assays Enzyme Assays
SensoLyte® Luminescent Alkaline Phosphatase Assay Kit provides highly sensitive chemiluminescent substrate to quantify alkaline phosphatase activity in solutions, in cell extracts, in live cells.
Phosphatases are important classes of enzymes involved in cellular signal transduction, cell metabolism and other biological activities. Changes in alkaline phosphatase level and activity are involved in a variety of physiological and pathological events, such as bone development, bone-related diseases, gestation related diseases, inflammatory bowel disease, post-parathyroidectomy stage and drug toxicity. Phosphatases have attracted considerable interest as: Enzyme markers, Drug screening targets, Conjugating agents and Reporter for gene expression studies.
Features: Kits include all essential components (chromogenic/fluorogenic/luminometry substrates) and sample reagents needed to perform 500 assays using 96-well plate format.
Optimized to detect specific alkaline phosphatase activity; compatible for HTS; detailed protocol and references provided.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty