Would you like to log in again?
SensoLyte® AFC Plasmin Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72124
103010-456EA 641.62 USD InStock
103010-456
SensoLyte® AFC Plasmin Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
Plasmin belongs to the family of serine proteases
The SensoLyte® AFC Plasmin Activity Assay Kit provides a convenient assay for high throughput screening of plasmin inhibitors and inducers or for continuous assay of plasmin activity. The assay utilizes a fluorogenic peptide, which upon enzyme cleavage releases the AFC (7-amido-4-trifluoromethylcoumarin) fluorophore. The fluorescent signal can be measured at Ex/ Em =380 nm/500 nm. This assay can detect picogram level of plasmin activity. Assays are performed in a convenient 96-well microplate format.
It plays a key role in fibrinolysis by dissolving fibrin in blood clots. Besides fibrinolysis, plasmin is also involved in such physiological and pathological processes as wound healing, liver repair, and the maintenance of liver homeostasis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty