Would you like to log in again?
SensoLyte® Calcein Cell Viability Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72126
103010-460EA 507.15 USD InStock
103010-460
SensoLyte® Calcein Cell Viability Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
Calcein AM (calcein acetoxymethyl ester) is a non-fluorescent reagent that easily permeates cellular membrane
The SensoLyte® Calcein Cell Viability Assay Kit provides a convenient tool to detect live cells. This kit can be used in proliferation, cytotoxicity, invasion, adhesion, migration and other cell-based assays.
Once it permeates live cells, esterases remove the acetomethoxy group allowing calcein to bind to calcium, which results in a strong green fluorescence. The increase of fluorescence is proportional to the number of live cells. Inside dead cells, the non-fluorescent calcein AM remains intact due to the lack of active esterases. The fluorescent signal from the conversion of calcein AM to calcein can be monitored at Ex/Em=494nm /520 nm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty