Would you like to log in again?
SensoLyte® Luminescent Peroxidase Assay Kit (Luminometric), AnaSpec Inc.
Supplier: AnaSpec
AS-72127
103010-462EA 631.38 USD InStock
103010-462
SensoLyte® Luminescent Peroxidase Assay Kit (Luminometric), AnaSpec Inc.
Assays Enzyme Assays
Peroxidases catalyze oxidation-reduction reactions and play an important role in protecting cell from oxidative injury
The SensoLyte® Luminescent Peroxidase Assay Kit provides highly sensitive chemiluminescent substrate to quantify peroxidase activity in solutions, in cell extracts, in live cells. The kit contains: chemiluminescent substrate, positive control (horseradish peroxidase), assay buffer, and an optimized ‘mix and read’ assay protocol that is compatible with HTS liquid handling instruments.
HRP-protein conjugates are extensively used as secondary detection reagents in ELISA, immunohistochemistry and assay development.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty