Would you like to log in again?
SensoLyte® 520 Thrombin Activity Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72129
103010-466EA 931 USD InStock
103010-466
SensoLyte® 520 Thrombin Activity Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
Thrombin, a serine protease, plays a central role in hemostasis by converting soluble plasma fibrinogen into an insoluble fibrin clot and by promoting platelet aggregation
The SensoLyte® 520 Thrombin Assay Kit is optimized for detecting thrombin activity. This kit contains a novel internally quenched 5-FAM/QXL® 520 FRET substrate for thrombin. Thrombin cleaves the FRET substrate into two separate fragments resulting in the release of 5-FAM fluorescence which can be monitored at excitation/emission= 490 nm/520 nm. The long wavelength fluorescence of 5-FAM is less interfered by the autofluorescence of components in biological samples and test compounds. The assay can be performed in a convenient 96-well or 384-well microplate format for screening of enzyme inhibitors.
Thrombin also influences a number of normal and pathological processes, including inflammation, tissue repair, embryogenesis, angiogenesis, and tumor invasion.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty