Would you like to log in again?
SensoLyte® AFC Thrombin Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72130
103010-468EA 808.23 USD InStock
103010-468
SensoLyte® AFC Thrombin Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
Thrombin, a serine protease, plays a central role in hemostasis by converting soluble plasma fibrinogen into an insoluble fibrin clot and by promoting platelet aggregation
The SensoLyte® AFC Thrombin Assay Kit is optimized for screening of enzyme inhibitors. This kit contains a fluorogenic substrate which shows high reactivity and low background. Thrombin cleaves the substrate resulting in the release of the AFC (7-amido-4-trifluoromethylcoumarin) fluorophore. Fluorescence can be monitored at excitation/emission= 380 nm/500 nm. The assays are performed in a convenient 96-well or 384-well microplate format.
Thrombin also influences a number of normal and pathological processes, including inflammation, tissue repair, embryogenesis, angiogenesis, and tumor invasion.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty