Would you like to log in again?
SensoLyte® FDG β-Galactosidase Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72133
103010-472EA 532 USD InStock
103010-472
SensoLyte® FDG β-Galactosidase Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
β-galactosidase, encoded by the lacZ gene in E
The SensoLyte® FDG β-Galactosidase Assay Kit provides a sensitive fluorogenic substrate for detecting β-galactosidase activity in tranfected cells. In the presence of β-galactosidase, the colorless fluorescein di-β-D galactopyranoside (FDG) substrate is hydrolyzed into a fluorescein that can be detected at Ex/Em = 490/520 nm.
coli, is widely used as a reporter enzyme to study gene expression, protein-protein interaction and normalization of transfection efficiency in mammalian cells.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty