Would you like to log in again?
SensoLyte® ONPG β-Galactosidase Assay Kit (Colorimetric), AnaSpec
Supplier: AnaSpec
AS-72134
103010-474EA 391.69 USD InStock
103010-474
SensoLyte® ONPG β-Galactosidase Assay Kit (Colorimetric), AnaSpec
Assays Enzyme Assays
β-galactosidase, encoded by the lacZ gene in <italic>E. coli</italic>, is widely used as a reporter enzyme to study gene expression, protein-protein interaction and normalization of transfection efficiency in mammalian cells.
The SensoLyte® ONPG β-Galactosidase Assay Kit utilizes the widely used chromogenic substrate, o-nitrophenyl β-D-galactopyranoside (ONPG) to determine the activity of β-galactosidase in transfected cells or tissues. In the presence of β-galactosidase, the colorless compound is hydrolyzed into o-nitrophenol, producing a yellow color that can be detected at absorbance 420 nm. The intensity of the color produced is proportional to β-galactosidase activity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty