Would you like to log in again?
SensoLyte® Thiol Quantitation Assay Kit (Colorimetric), AnaSpec
Supplier: AnaSpec
AS-72136
103010-476EA 507.15 USD InStock
103010-476
SensoLyte® Thiol Quantitation Assay Kit (Colorimetric), AnaSpec
Assays Enzyme Assays
Thiols, compounds that contain sulfhydryl group, are powerful reducing agents capable of acting as antioxidants, enzyme cofactors, and neuromodulators.
The SensoLyte® Thiol Quantitation Assay Kit utilizes the widely used Elmann’s reagent for colorimetric measurement of thiol concentrations. The sulfhydryl group of thiol reacts with Ellman’s Reagent [5, 5’-dithiobis (2-nitrobenzoic acid)] and generates 2-nitro-5-thiobenzoic acid (TNB) producing a yellow color that can be detected at absorbance 420 nm. The intensity of the color produced is proportional to thiol concentration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty