Would you like to log in again?
SensoLyte® ABD-F Thiol Quantitation Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72137
103010-478EA 600.69 USD InStock
103010-478
SensoLyte® ABD-F Thiol Quantitation Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
Thiols, compounds that contain sulfhydryl group, are powerful reducing agents capable of acting as antioxidants, enzyme cofactors, and neuromodulators.
The SensoLyte® ABD-F Thiol Quantitation Assay Kit provides a convenient, sensitive fluorescent assay for measurement of thiol concentrations. 4-fluoro-7-aminosulfonylbenzofurazan (ABD-F), the reagent provided in the kit, generates yellow-green fluorescence upon reaction with thiols. Fluorescence can be monitored at excitation/emission=389 nm/513 nm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty