Would you like to log in again?
SensoLyte® 520 Thiol Quantitation Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72138
103010-480EA 733.69 USD InStock
103010-480
SensoLyte® 520 Thiol Quantitation Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
Thiols, compounds that contain sulfhydryl group, are powerful reducing agents capable of acting as antioxidants, enzyme cofactors, and neuromodulators.
The SensoLyte® 520 Thiol Quantitation Assay Kit provides a convenient, highly sensitive fluorescent assay for measurement of thiol concentrations. The novel reagent supplied in the kit, upon reaction with thiols, generates fluorescence which can be easily detected at excitation/emission=490/520 nm. The assay has a linear range of 0.04 to 160 µM thiol.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty