Would you like to log in again?
SensoLyte® 520 Rat Renin Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72140
103010-482EA 1140 USD InStock
103010-482
SensoLyte® 520 Rat Renin Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
Renin, a highly specific aspartyl protease, cleaves angiotensinogen, to yield angiotensin I, which is further converted into angiotensin II by ACE (Angiotensin Converting Enzyme)
The SensoLyte® 520 Rat Renin Assay Kit provides a convenient assay for high throughput screening of renin inhibitors and for continuous assay of renin activity using a 5-FAM/QXL®520 fluorescence resonance energy transfer (FRET) peptide. In the FRET peptide the fluorescence of 5-FAM is quenched by QXL®520. Upon cleavage into two separate fragments by renin, the fluorescence of 5-FAM is recovered, and can be monitored at excitation/emission = 490/520 nm.
Angiotensin II constricts blood vessels leading to increased blood pressure. Discovery of new renin inhibitors for treatment of hypertension is a promising area of research.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty