Would you like to log in again?
SensoLyte® pNPP Secreted Alkaline Phosphatase Reporter Gene Assay Kit Colorimetric, AnaSpec, Inc.
Supplier: AnaSpec
AS-72144
103010-490EA 339.08 USD InStock
103010-490
SensoLyte® pNPP Secreted Alkaline Phosphatase Reporter Gene Assay Kit Colorimetric, AnaSpec, Inc.
Assays Enzyme Assays
The secreted alkaline phosphatase (SEAP) is widely used as reporter gene to analyze gene expression including kinetic analysis over time in cell culture or animals owing to its unique ability to secrete into culture medium and serum
  • Features: Kits include all essential components (chromogenic/fluorogenic/luminometry substrates) and sample reagents needed to perform 500 assays using 96-well plate format.
  • Optimized to detect specific alkaline phosphatase activity; compatible for HTS; detailed protocol and references provided.
Phosphatases are important classes of enzymes involved in cellular signal transduction, cell metabolism and other biological activities. Changes in alkaline phosphatase level and activity are involved in a variety of physiological and pathological events, such as bone development, bone-related diseases, gestation related diseases, inflammatory bowel disease, post-parathyroidectomy stage and drug toxicity. Phosphatases have attracted considerable interest as: Enzyme markers, Drug screening targets, Conjugating agents and Reporter for gene expression studies.
SensoLyte® pNPP Secreted Alkaline Phosphatase Reporter Gene Assay Kit provides a sensitive colorimetric assay of placental alkaline phosphatase for both secreted and membrane-bound form of enzyme
Ordering information: Kits include all essential components (chromogenic/fluorogenic/luminometry substrates) and sample reagents needed to perform 500 assays using 96-well plate format.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty