Would you like to log in again?
SensoLyte® AMC Calpain Activity Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72150
103010-506EA 898.85 USD InStock
103010-506
SensoLyte® AMC Calpain Activity Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
The calpains are a family of intracellular Ca2+ supdependent cysteine proteases
The SensoLyte® AMC Calpain Assay Kit is optimized for detecting calpain activities. This kit contains a fluorogenic peptide substrate, suc-LLVY-AMC. Upon calpain protease cleavage, suc-LLVY-AMC generates the AMC fluorophore emitting bright blue fluorescence and can be monitored at excitation/emission=354/442 nm. Increase in AMC fluorescence is proportional to calpain activity. The assay can detect both calpain 1 (μm) and 2 (m) activities and is ideal for kinetic study of these enzymes. The assays are performed in a convenient 96-well microplate format.
The best-characterized calpains are isoforms 1 (µm) and 2 (m). Calpains respond to Ca2+ signals by cleaving many specific proteins, thereby irreversibly modifying their function(s). They are implicated in a variety of Ca2+ regulated cellular processes as well as various pathological phenomena, such as ischemic injury, muscular dystrophy, diabetes, cataract, atherosclerosis, Alzheimer's disease, and cancer. Calpains represent potential therapeutic targets for drug discovery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty