Would you like to log in again?
SensoLyte® Rh110 Cathepsin K Assay Kit Fluorimetric, AnaSpec, Inc.
Supplier: AnaSpec
AS-72152
103010-508EA 887.15 USD InStock
103010-508
SensoLyte® Rh110 Cathepsin K Assay Kit Fluorimetric, AnaSpec, Inc.
Assays Enzyme Assays
Cathepsin K is the lysosomal cysteine protease involved in bone remodeling and resorption
Cathepsins are a class of globular lysosomal proteases playing a vital role in mammalian cellular turnover. They degrade polypeptides and are distinguished by their substrate specificities.
The cathepsin assay kits offers a series of fluorimetric long wavelength, sensitive SensoLyte® 520 assay kits for enzyme activity determination of Cathepsins G, D, E, B, K, L and S. These kits employ a FRET (Fluorescence Resonance Energy Transfer) peptide substrate that emits in the green range of 520 nm, with excitation at 490 nm. The advantage of using longer wavelength emitting substrates means that autofluorescence from cellular components and test compounds is minimal; thus making the kits more sensitive. Furthermore, the use of HiLyte Fluor™ 488 (except for Cathepsin S, which uses 5-FAM) as the FRET donor (fluorophore) results in a more stable fluorescent signal, since HiLyte Fluor™ 488 is resistant to acidic pH. A listing of the SensoLyte® Cathepsin Kits providing conventional FRET (Mca/Dnp) or fluorogenic substrates for cathepsins are also available.
Kits detect specific Cathepsin activities; provides ample reagents to perform 100 assays in 96-well format; minimal hands-on time required; detailed protocol and references provided.
It has potential as a drug target in autoimmune diseases and osteoporosis. The SensoLyte® Rh110 Cathepsin K Assay Kit provides a convenient assay for high throughput screening of enzyme inhibitors or for continuous assay of enzyme activity utilizing a fluorogenic peptide. Upon enzyme cleavage, the peptide releases the Rh110 fluorophore with bright green fluorescence that can be detected at Ex/Em=496/520 nm.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty