Would you like to log in again?
SensoLyte® 520 Total GSH Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72154
103010-512EA 770.23 USD InStock
103010-512
SensoLyte® 520 Total GSH Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
Glutathione (GSH) is an important antioxidant molecule that functions as a cofactor for a variety of enzymes and is involved in various biological processes
The SensoLyte® 520 Total GSH Quantitation Assay Kit utilizes an optimized enzymatic recycling method for the quantification of glutathione. Glutathione reductase converts oxidized glutathione to reduced glutathione. Upon reaction with reduced GSH, the novel proprietary thiol detection reagent supplied in the kit generates fluorescence which can be easily detected at excitation/emission =490/520 nm. The assay has a linear range of 0.2 to 100 µM GSH. The kit also provides 5-sulfosalicylic acid for protein removal from biological samples.
Total GSH includes reduced GSH and oxidized GSH (GSSG). The concentrations of GSH and GSSG and their molar ratio can be used for indicating the oxidative stress status.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty