Would you like to log in again?
SensoLyte® 520 FRET SIRT1 Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72155
103010-514EA 827.23 USD InStock
103010-514
SensoLyte® 520 FRET SIRT1 Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
Histone deacetylase (HDAC) enzymes act as transcriptional repressors of genes through the deacetylation of lysine residues on histone proteins
The SensoLyte® 520 FRET SIRT1 Assay Kit provides a convenient, two-step homogeneous procedure for measuring sirtuin 1 activity using FRET substrate. In the first step, an acetylated substrate is incubated with SIRT1 containing samples. Deacetylation of substrate first sensitizes it, so that, in the second step mixing with the SIRT developer generates a fluorophore, which can then be detected at Ex/Em= 490/520 nm. This kit can be used for high throughput screening of SIRT1 inhibitors/activators. The long wavelength fluorescence of FRET substrate is less interfered by the autofluorescence of cell components and test compounds. The nuclear extract HDAC (containing HDACs 1 and 2) does not deacetylate FRET sirtuin substrate. Assays are performed in a standard 96-well format.
Sirtuins (SIRTs) are a unique class of deacetylases (class III HDACs) that require NAD as a cofactor. Sirtuin 1 (SIRT1) is involved in regulation of tumor suppressor p53 and inhibition of apoptosis. It also represents a target for treatment of age-related diseases and type II diabetes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty