Would you like to log in again?
SensoLyte® GST Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72157
103010-518EA 552.46 USD InStock
103010-518
SensoLyte® GST Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
GST (Glutathione S-Transferase) enzymes, a family of isozymes plays a crucial role in body defense mechanisms against toxins and carcinogens
The SensoLyte® GST Activity Assay Kit utilizes an optimized method for detection of enzyme activity in biological samples. The kit provides substrate for GST enzyme, which upon reaction with reduced glutathione yields a fluorescent glutathione adduct. This produces bright blue fluorescence that can be monitored at excitation/emission =390 nm/480 nm.
GSTs carry out their functions mostly by mediating their conjugation with reduced glutathione on substrates such as pharmaceutical and environmental pollutants.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty