Would you like to log in again?
SensoLyte® Glutathione Cellular Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72158
103010-520EA 552.46 USD InStock
103010-520
SensoLyte® Glutathione Cellular Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
Glutathione (GSH) is an important antioxidant molecule that functions as a cofactor for a variety of enzymes and is involved in various biological processes
The SensoLyte® Glutathione Cellular Assay Kit utilizes an optimized enzymatic method for determination of the glutathione levels in cell or tissue extracts. The substrate provided in the kit can selectively react with GSH in the reaction catalyzed by glutathione S-transferase (GST) and form a fluorescent glutathione adduct. The resulting blue fluorescence can be monitored at excitation /emission= 390 nm/480 nm. The assay has a linear range of 0-100 µM of GSH. This kit does not require the usage of deproteination for biological samples.
A variation in glutathione levels is an indicator of oxidative stress.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty