Would you like to log in again?
SensoLyte® AFC Urokinase (uPA) Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72159
103010-522EA 825.77 USD InStock
103010-522
SensoLyte® AFC Urokinase (uPA) Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
Urokinase-type plasminogen activator (uPA) is a serine protease that functions in the conversion of the zymogen plasminogen to the active, broad-spectrum serine protease plasmin
The SensoLyte® AFC Urokinase (uPA) Assay Kit is optimized for screening of enzyme inhibitors. This kit contains a fluorogenic substrate with a high reactivity and low background. uPA cleaves the substrate resulting in release of AFC (7-amino-4-trifluoromethylcoumarin) fluorophore. Fluorescence can be monitored at excitation /emission= 380/500 nm.
Plasmin, in turn, mediates the pericellular proteolysis of extracellular matrix components and activates other proteases such as matrix metalloproteinases and collagenases that lead to the further degradation and remodeling of the extracellular matrix. Besides the proteolytic function, upon binding to uPA receptor (uPAR), uPA is involved in initiating versatile intracellular signal pathways that regulate cell proliferation, adhesion, and migration through its interaction with various integrins and vitronectin. uPA has been implicated in a number of normal and pathological processes, and recognized as a prognostic marker for various cancers as well as a target for drug discovery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty