Would you like to log in again?
SensoLyte® HAT (p300) Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72172
103010-548EA 893 USD InStock
103010-548
SensoLyte® HAT (p300) Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
Histone acetyltransferases (HATs) enzymes regulate the acetylation of histones and non-histone proteins
The SensoLyte® HAT (p300) Assay Kit provides a convenient protocol for screening of enzyme inhibitors and for continuous assay of p300 activity. For added convenience, this kit includes two peptide substrates: histone H3 (1-21) peptide and non-histone p53 peptide (368-386). After incubation with acetyl CoA and the substrate, the p300 enzyme generates acetylated H3 or p53 peptide and CoASH. The thiol groups of CoASH can be detected with fluorogenic reagent at excitation/ emission=389nm/513nm.
The acetylation of the e-amino groups of lysine residues present at histone tails correlates largely with transcriptional activation, but it is also involved in DNA replication, DNA repair and protein–protein interactions. HATs play major roles in the control of cell fate and their misregulation is implicated in the development of some human tumors. HAT p300 is a transcriptional coactivator that acetylates core histones facilitating chromatin decondensation and recruiting basic RNA polymerase machinery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty