Would you like to log in again?
SensoLyte® Green Elastase Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72178
103010-566EA 507.15 USD InStock
103010-566
SensoLyte® Green Elastase Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
The SensoLyte® Green Elastase Assay Kit is a FRET-based assay that detects elastase activity. The kit provides an elastin, natural substrate for elastases, labeled with 5-FAM fluorophore and QXL®520 quencher. Elastase-catalyzed hydrolysis yields brightly green fluorescence. Increase in fluorescence intensity is directly proportional to enzyme activity. The kit does not require any separation steps and can be used to continuously measure the kinetics of elastase.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty