Would you like to log in again?
SensoLyte® Rh110 Elastase Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72179
103010-568EA 507.15 USD InStock
103010-568
SensoLyte® Rh110 Elastase Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
The SensoLyte® Rh110 Elastase Assay Kit detects elastase activity using a fluorogenic peptide substrate. Upon elastase cleavage, the peptide releases the Rh110 fluorophore with bright green fluorescence that can be detected at Ex/Em=496 /520 nm. The increase in fluorescence intensity is directly proportional to enzyme activity. The kit does not require any separation steps and can be used to continuously measure the kinetics of elastase. The longer-wavelength spectra and higher extinction coefficient of the Rh110 provide greater sensitivity and less interference from screening compounds.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty