Would you like to log in again?
SensoLyte® 520 Cathepsin G Assay Kit Fluorimetric, AnaSpec, Inc.
Supplier: AnaSpec
AS-72185
103010-580EA 893 USD InStock
103010-580
SensoLyte® 520 Cathepsin G Assay Kit Fluorimetric, AnaSpec, Inc.
Assays Enzyme Assays
Cathepsin G is the serine protease released by neutrophils upon their activation
Cathepsins are a class of globular lysosomal proteases playing a vital role in mammalian cellular turnover. They degrade polypeptides and are distinguished by their substrate specificities.
The cathepsin assay kits offers a series of fluorimetric long wavelength, sensitive SensoLyte® 520 assay kits for enzyme activity determination of Cathepsins G, D, E, B, K, L and S. These kits employ a FRET (Fluorescence Resonance Energy Transfer) peptide substrate that emits in the green range of 520 nm, with excitation at 490 nm. The advantage of using longer wavelength emitting substrates means that autofluorescence from cellular components and test compounds is minimal; thus making the kits more sensitive. Furthermore, the use of HiLyte Fluor™ 488 (except for Cathepsin S, which uses 5-FAM) as the FRET donor (fluorophore) results in a more stable fluorescent signal, since HiLyte Fluor™ 488 is resistant to acidic pH. A listing of the SensoLyte® Cathepsin Kits providing conventional FRET (Mca/Dnp) or fluorogenic substrates for cathepsins are also available.
Kits detect specific Cathepsin activities; provides ample reagents to perform 100 assays in 96-well format; minimal hands-on time required; detailed protocol and references provided.
Cathepsin G is actively being explored as a target for anti-inflammatory and anti-infective drugs. The SensoLyte® 520 Cathepsin G Activity Assay Kit uses a HiLyte™ Fluor 488/QXL® 520 FRET peptide substrate for the measurement of enzyme activity. In the intact FRET peptide, the fluorescence of HiLyte™ Fluor 488 is quenched by QXL® 520. Upon cleavage of the FRET peptide by the active enzyme, the increase of fluorescence can be continuously monitored at excitation/emission = 490/520 nm. With superior fluorescence quantum yield and longer emission wavelength, the HiLyte™ Fluor 488/QXL® 520 based FRET peptide has less interference from the autofluorescence of test compounds and cellular components and provides better assay sensitivity. The fluorescent signal from HiLyte™ Fluor 488 is stable at low pH, which is the optimal pH for cathepsin activity. The kit can be used to detect the activity of Cathepsin G enzyme in biological samples and purified enzyme preparations.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty