Would you like to log in again?
Human Recombinant Sirtuin (from E. coli)
Supplier: AnaSpec
AS-72187
103010-584EA 197.31 USD InStock
103010-584
Human Recombinant Sirtuin (from E. coli)
Proteins and Peptides
This human recombinant SIRT2 is in its active form, and can be used for enzyme activity assays. The recombinant human Sirtuin 2 (GenBank Accession #: NM_030593) with 13-319 amino acids and His tag at its C-terminal was expressed in E. coli. The molecular mass of the enzyme is approximately 35.5 kDa on SDS-PAGE.
Sirtuins comprise a unique class of nicotinamide adenine dinucleotide (NAD+)-dependent deacetylases (class III HDACs) targeting multiple protein substrates.
Sirtuin 1 (SIRT1), the human homolog of yeast Sir2 (Silent Information Regulator 2), is the most studied of the seven members of sirtuin family. SIRT1 have been implicated in several important cellular processes, including genomic stability and DNA repair, p53-mediated apoptosis, adipogenesis, and aging.
Substrates for SIRT2 are not limited to histones but also include various transcription factors and co-regulators that modulate metabolic, cell cycle and cell death related pathways. Human SIRT2 is a cytoplasmic protein that increases in abundance during mitosis and regulates major events of cytokinesis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty