Would you like to log in again?
SensoLyte® Green SIRT2 Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72188
103010-586EA 893 USD InStock
103010-586
SensoLyte® Green SIRT2 Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
Histone deacetylase (HDAC) enzymes act as transcriptional repressors of genes through the deacetylation of lysine residues on histone proteins
The SensoLyte® Green SIRT2 Assay Kit provides a convenient, two-step homogeneous procedure for measuring sirtuin 2 activity using a fluorogenic substrate. In the first step, an acetylated substrate is incubated with SIRT2 containing samples. Deacetylation of substrate first sensitizes it, so that, in the second step, mixing with the SIRT developer generates a fluorophore, which can then be detected at Ex/Em= 490 nm/520 nm. This kit can be used for high throughput screening (HTS) of SIRT2 inhibitors/activators. The long wavelength fluorescence of SIRT2 substrate is less interfered by the autofluorescence of cell components and test compounds. Assays are performed in a standard 96-well plate format.
Sirtuins (SIRTs) are a unique class of deacetylases (class III HDACs) that require NAD+ as a cofactor. Sirtuin 2 (SIRT2) is a cytoplasmic protein involved in regulation of cell cycle, DNA repair and tumorogenesis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty