Would you like to log in again?
SensoLyte® Rh110 DPPIV Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72196
103010-600EA 667.92 USD InStock
103010-600
SensoLyte® Rh110 DPPIV Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
DPPIV is a widely distributed serine protease that cleaves two amino acids from small peptides containing alanine or proline in the second position of the N-terminus of the peptide
The SensoLyte® Rh110 DPPIV Assay Kit provides a convenient method for high throughput screening of DPPIV inhibitors and for continuous assay of DPPIV activity utilizing a fluorogenic peptide. Upon enzyme cleavage, the peptide releases the Rh110 fluorophore with bright green fluorescence that can be detected at Ex/Em=490/520 nm. The longer-wavelength spectra and higher extinction coefficient of the Rh110 provide greater sensitivity and less interference from reaction components.
DPPIV activity is especially critical for inactivation of GLP-1, a potent antihyperglycemic hormone, inducing glucose-dependent stimulation of insulin secretion. DPPIV inhibitors provide a potent treatment for type 2 diabetes prolonging the activity of GLP-1 and improving insulin secretion and blood glucose control regulation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty