Would you like to log in again?
SensoLyte® AMC DPPIV Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72197-
103010-602EA 552.46 USD InStock
103010-602
SensoLyte® AMC DPPIV Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
DPPIV is a widely distributed serine protease that cleaves two amino acids from small peptides containing alanine or proline in the second position of the N-terminus of the peptide
The SensoLyte® AMC DPPIV Assay Kit provides a convenient method for high throughput screening (HTS) of DPPIV inhibitors and for continuous assay of DPPIV activity using a fluorogenic substrate that has a high reactivity and low background. DPPIV cleaves the substrate resulting in release of AMC (7-amino-4-methylcoumarin) fluorophore. Fluorescence can be monitored at excitation /emission= 354/442 nm. This kit can be used for high throughput screening of DPPIV inhibitors.
DPPIV activity is especially critical for inactivation of GLP-1, a potent antihyperglycemic hormone, inducing glucose-dependent stimulation of insulin secretion. DPPIV inhibitors provide a potent treatment for type 2 diabetes prolonging the activity of GLP-1 and improving insulin secretion and blood glucose control regulation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty