Would you like to log in again?
SensoLyte® 390 Generic MMP Activity Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72202
103010-612EA 698.62 USD InStock
103010-612
SensoLyte® 390 Generic MMP Activity Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated proteins capable of digesting extracellular matrix components
The SensoLyte® 390 Generic MMP Assay Kit is optimized to detect the activity of a variety of MMPs, including MMP-1, 2, 7, 8, 9, 13, 14, 15, 16, and 24. This kit can be used for detecting generic MMP activity in biological samples or for high throughput screening of MMP inducers and inhibitors using purified MMPs. This kit provides an Mca/Dnp fluorescence resonance energy transfer (FRET) peptide as a MMP substrate. In the intact FRET peptide, the fluorescence of Mca is quenched by Dnp. Upon cleavage into two separate fragments by MMPs, the fluorescence of Mca is recovered, and can be monitored at excitation/emission wavelengths = 330 nm/390 nm. The assays are performed in a convenient 96-well microplate format.
MMPs play important roles in tumor development and invasion, as well in many diseases.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty