Would you like to log in again?
SensoLyte® 520 Deubiquitination Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72203
103010-614EA 735.15 USD InStock
103010-614
SensoLyte® 520 Deubiquitination Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
Deubiquitinating enzymes (DUBs) are proteases that reverse ubiquitin modifications and can rescue a protein from proteasomal degradation
The SensoLyte® 520 Deubiquitination Assay Kit provides a convenient assay for the screening of enzyme inhibitors and activators or for continuous assay of deubiquitination activity through the use of a fluorogenic peptide substrate. Upon the cleavage by a deubiquitin protease, this substrate generates the green fluorophore that can be detected at excitation/emission=490/520 nm.
DUBs specifically cleave ubiquitin-linked molecules downstream of the terminal carbonyl of the last residue of ubiquitin (Gly76). They are classified in five families. Well-defined families include ubiquitin carboxyl-terminal hydrolases (UCH) and ubiquitin-processing proteases (UBP). DUBs act as regulators of diverse cellular processes, such as development, growth and DNA repair. They have become actively studied molecular targets for drug discovery since they are implicated in the pathogenesis of many human diseases.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty