Would you like to log in again?
SensoLyte® 440 Deubiquitination Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72204
103010-616EA 967.54 USD InStock
103010-616
SensoLyte® 440 Deubiquitination Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
Deubiquitinating enzymes (DUBs) are proteases that reverse ubiquitin modifications and can rescue a protein from proteasomal degradation
The SensoLyte® 440 Deubiquitination Assay Kit provides a convenient assay for the screening of enzyme inhibitors and activators or for continuous assay of deubiquitination activity. A natural substrate, ubiquitin, coupled with the AMC (7-amino-4-methylcoumarin) fluorophore is used in the assay. Upon the cleavage by a deubiquitin protease, this substrate generates the blue fluorophore that can be detected at excitation/emission=354/442 nm.
They specifically cleave ubiquitin-linked molecules downstream of the terminal carbonyl of the last residue of ubiquitin (Gly76). DUBs are classified into five families. Well-defined families include ubiquitin carboxyl-terminal hydrolases (UCH) and ubiquitin-processing proteases (UBP). They act as regulators of diverse cellular processes, such as development, growth and DNA repair. DUBs have become actively studied molecular targets for drug discovery since they are implicated in the pathogenesis of many human diseases.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty