Would you like to log in again?
SensoLyte® NADP/NADPH Assay Kit (Colorimetric), AnaSpec
Supplier: AnaSpec
AS-72205
103010-618EA 697.15 USD InStock
103010-618
SensoLyte® NADP/NADPH Assay Kit (Colorimetric), AnaSpec
Assays Enzyme Assays
Nicotinamide nucleotides are key players in the energy transforming and oxidation-reduction reactions of a cell
The SensoLyte® NADP/NADPH Assay Kit utilizes an optimized enzymatic recycling method for the quantification of NADP/NADPH without interfering with NAD/NADH. The final product of reaction, formazan, has blue color and can be detected at absorbance 565 nm. The intensity of the color produced is proportional to NADP/NADPH concentration.
NADP and NADPH act as allosteric effectors in modulating enzyme activities. The reducing power of NADPH is a required cofactor for enzymes that are pro-oxidants such as nitric oxide synthase and NADPH oxidase. NADP/NADPH ratio is involved in the control of the oxidative shunt. Each of these forms has a particular role during differentiation and similarity processes as well as being target functions in the cytotoxic action of ionizing radiations and alkylating agents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty